5-Methylisoxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A273491-100G A273491-10G A273491-25G A273491-500G A273491-5G
77014-466EA 91.24 USD
77014-466 77014-468 77014-470 77014-472 77014-474
5-Methylisoxazole-4-carboxylic acid 97%
5-Methylisoxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.1 g/mol
MDL Number: MFCD00955655
CAS Number: 42831-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR