4-Bromo-7-fluoro-1H-indazole 98%, Ambeed.Inc

Supplier: Ambeed
Danger

A273456-100MG A273456-250MG A273456-1G A273456-25G A273456-5G
77014-462EA 11 USD
77014-462 77014-464 77683-831 77892-534 77892-535
4-Bromo-7-fluoro-1H-indazole 98%, Ambeed.Inc
4-Bromo-7-fluoro-1H-indazole

Precaución: For research use only.

Formula: C₇H₄BrFN₂
MDL Number: MFCD16658655
CAS Number: 1186334-63-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR