Rosuvastatin calcium salt 98%

Supplier: Ambeed
Danger

Synonyms: 7-{4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulphonyl)amino]pyrimidin-5-yl}-3,5-dihydroxyhept-6-enoic acid, Calcium (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulphonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate, Rosuvastatin Calcium, CRESTOR, (3R,5S,6E)-7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[N-methyl(n-methylsulphonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-Heptenoic calcium

A272688-10MG A272688-50MG A272688-5MG A272688-1G A272688-25G A272688-5G
77001-620EA 8 USD
77001-620 77001-622 77001-624 77571-682 77601-172 77601-174
Rosuvastatin calcium salt 98%
Rosuvastatin calcium salt

Precaución: for Research Use Only

Formula: C₂₂H₂₇FN₃O₆S·¹/₂Ca
MW: 501.58 g/mol
MDL Number: MFCD04112702
CAS Number: 147098-20-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR