Dimethyl-L-aspartate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Aminosuccinic Acid Dimethyl Ester Hydrochloride, L-Asparaginic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-Aminosuccinate Hydrochloride, H-Asp(ome)-OMe HCl, Dimethyl L-Asparaginate Hydrochloride, H-Asp(OMe)-OMe.HCl, L-Aspartic Acid Dimethyl Ester Hydrochloride
A272538-10G
A272538-5G
A272538-100G
77001-610EA
10.61
USD
77001-610
77001-612
77672-408
Dimethyl-L-aspartate hydrochloride 98%
Dimethyl-L-aspartate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₄ MW: 197.62 g/mol |
MDL Number:
MFCD00038878 CAS Number: 32213-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...