6-Fluoroimidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A272219-250MG A272219-100MG
77001-604EA 36.08 USD
77001-604 77672-404
6-Fluoroimidazo[1,2-a]pyridine 95%
6-Fluoroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1300 g/mol
MDL Number: MFCD09994632
CAS Number: 139022-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR