4-Bromo-6-fluoro-1H-indazole 95%
Supplier: Ambeed
|
Warning
|
A272048-100MG
A272048-1G
A272048-250MG
A272048-25G
A272048-5G
77001-590EA
6
USD
77001-590
77001-592
77001-594
77001-596
77672-402
4-Bromo-6-fluoro-1H-indazole 95%
4-Bromo-6-fluoro-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄BrFN₂ |
MDL Number:
MFCD07781448 CAS Number: 885520-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...