(S)-1-Boc-3-(Bromomethyl)pyrrolidine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A271799-250MG A271799-100MG A271799-1G
76988-320EA 122.01 USD
76988-320 77683-828 77683-829
(S)-1-Boc-3-(Bromomethyl)pyrrolidine 98%, Ambeed.Inc
(S)-1-Boc-3-(Bromomethyl)pyrrolidine

Precaución: For research use only.

Formula: C₁₀H₁₈BrNO₂
MW: 264.16 g/mol
MDL Number: MFCD08059334
CAS Number: 1067230-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR