4-(Dimethoxymethyl)-N-methylpyrimidin-2-amine 95%

Supplier: Ambeed
Warning

A271754-100MG A271754-1G A271754-250MG A271754-5G
77014-426EA 33.95 USD
77014-426 77014-428 77014-430 77014-432
4-(Dimethoxymethyl)-N-methylpyrimidin-2-amine 95%
4-(Dimethoxymethyl)-N-methylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O₂
MW: 183.21 g/mol
MDL Number: MFCD10566061
CAS Number: 180869-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR