2-Methyl-1,3-benzothiazole-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methylbenzo[d]thiazole-5-carboxylic acid
A271483-250MG
A271483-100MG
A271483-1G
A271483-5G
77001-198EA
20.16
USD
77001-198
77601-158
77601-160
77672-400
2-Methyl-1,3-benzothiazole-5-carboxylic acid 95%
2-Methyl-1,3-benzothiazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₂S MW: 193.222 g/mol |
MDL Number:
MFCD00498510 CAS Number: 24851-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...