11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one 98%

Supplier: Ambeed
Warning

Synonyms: 11-(2-Chloroacetyl)-5H-benzo[e]pyrido[3,2-b][1,4]diazepin-6(11H)-one

A271444-100G A271444-1G A271444-25G
77001-188EA 101.85 USD
77001-188 77001-190 77001-192
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one 98%
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClN₃O₂
MW: 287.703 g/mol
MDL Number: MFCD21648226
CAS Number: 28797-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR