Isoquinoline-8-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 8-Cyano-2-azanaphthalene, 8-Cyanoisoquinoline

A271433-100MG A271433-1G A271433-250MG A271433-5G
77031-952EA 23.34 USD
77031-952 77031-954 77031-956 77031-958
Isoquinoline-8-carbonitrile 98%
Isoquinoline-8-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.168 g/mol
MDL Number: MFCD10699232
CAS Number: 362606-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR