4-Bromo-3-(trifluoromethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

A271367-100MG A271367-1G A271367-250MG
76988-314EA 98.67 USD
76988-314 76988-316 76988-318
4-Bromo-3-(trifluoromethyl)-1H-indazole 95%
4-Bromo-3-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD22200789
CAS Number: 1211583-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR