4-Cyano-4'-pentyloxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile

A271031-10G A271031-1G A271031-5G
77031-946EA 55 USD
77031-946 77031-948 77031-950
4-Cyano-4'-pentyloxybiphenyl 98%
4-Cyano-4'-pentyloxybiphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO
MDL Number: MFCD00074879
CAS Number: 52364-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR