2-[(2-Fluorophenyl)amino]nitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-2'-nitrodiphenylamine, 2-Fluoro-N-(2-nitrophenyl)aniline

A271021-1G A271021-25G A271021-5G
77001-182EA 18.04 USD
77001-182 77001-184 77001-186
2-[(2-Fluorophenyl)amino]nitrobenzene 95%
2-[(2-Fluorophenyl)amino]nitrobenzene

Precaución: for Research Use Only

Formula: C₁₂H₉FN₂O₂
MW: 232.21 g/mol
MDL Number: MFCD08741396
CAS Number: 28898-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR