4-(Pyridin-3-yloxy)benzoic acid 95%

Supplier: Ambeed
Warning

A270851-100MG A270851-1G A270851-250MG A270851-5G
77031-938EA 35.01 USD
77031-938 77031-940 77031-942 77031-944
4-(Pyridin-3-yloxy)benzoic acid 95%
4-(Pyridin-3-yloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₃
MW: 215.2100 g/mol
MDL Number: MFCD11182951
CAS Number: 437383-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR