1-Benzyl-3-phenoxypiperidine 97%

Supplier: Ambeed
Warning

A270850-100MG A270850-1G A270850-250MG
77031-932EA 22 USD
77031-932 77031-934 77031-936
1-Benzyl-3-phenoxypiperidine 97%
1-Benzyl-3-phenoxypiperidine

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO
MW: 267.37 g/mol
MDL Number: MFCD16659836
CAS Number: 136421-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR