tert-Butyl (5-bromo-3-methylpyridin-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A270681-100MG A270681-1G A270681-250MG A270681-5G A270681-25G
77031-924EA 5 USD
77031-924 77031-926 77031-928 77031-930 77872-318
tert-Butyl (5-bromo-3-methylpyridin-2-yl)carbamate 97%
tert-Butyl (5-bromo-3-methylpyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O₂
MDL Number: MFCD11109816
CAS Number: 748812-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR