3-Bromo-5-methoxyphenol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Brom-5-hydroxyanisole

A269802-100MG A269802-250MG A269802-1G
76988-310EA 29.71 USD
76988-310 76988-312 77550-682
3-Bromo-5-methoxyphenol 97%
3-Bromo-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
MDL Number: MFCD12405420
CAS Number: 855400-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR