5-Bromo-2-chloronicotinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloro-3-cyanopyridine

A269775-100G A269775-10G A269775-1G A269775-25G A269775-5G
77031-888EA 265.23 USD
77031-888 77031-890 77031-892 77031-894 77031-896
5-Bromo-2-chloronicotinonitrile 97%
5-Bromo-2-chloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MDL Number: MFCD09801046
CAS Number: 405224-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR