2,5-Diaminopyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A269535-100MG A269535-250MG
77014-402EA 28.65 USD
77014-402 77014-404
2,5-Diaminopyrimidin-4(3H)-one 95%
2,5-Diaminopyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₄H₆N₄O
MW: 126.12 g/mol
MDL Number: MFCD18427067
CAS Number: 77994-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR