2,6-Difluoro-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A269469-1G A269469-250MG A269469-25G A269469-5G
76988-302EA 28.65 USD
76988-302 76988-304 76988-306 76988-308
2,6-Difluoro-3-methoxybenzoic acid 98%
2,6-Difluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MDL Number: MFCD04115901
CAS Number: 886498-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR