2-Bromo-6-iodo-3-pyridinol 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-hydroxy-6-iodopyridine, 2-Bromo-6-iodopyridin-3-ol

A269346-10G A269346-250MG A269346-25G A269346-5G
77001-104EA 135.8 USD
77001-104 77001-106 77001-118 77001-120
2-Bromo-6-iodo-3-pyridinol 96%
2-Bromo-6-iodo-3-pyridinol

Precaución: for Research Use Only

Formula: C₅H₃BrINO
MW: 299.9 g/mol
MDL Number: MFCD09909342
CAS Number: 129611-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR