Methyl 2-(bromomethyl)-6-cyanobenzoate 97%
Supplier: Ambeed
|
Danger
|
A269334-100MG
A269334-1G
A269334-250MG
77014-396EA
29
USD
77014-396
77014-398
77014-400
Methyl 2-(bromomethyl)-6-cyanobenzoate 97%
Methyl 2-(bromomethyl)-6-cyanobenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₈BrNO₂ MW: 254.08 g/mol |
MDL Number:
MFCD18398836 CAS Number: 1022980-69-3 UN: 1759 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...