2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde oxime 98%

Supplier: Ambeed
Danger

A269239-100MG A269239-1G A269239-250MG
76988-288EA 15.92 USD
76988-288 76988-290 76988-292
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde oxime 98%
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde oxime

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.17 g/mol
MDL Number: MFCD00862517
CAS Number: 31127-39-6
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR