2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide 97%

Supplier: Ambeed
Warning

A269217-10G A269217-1G A269217-250MG A269217-25G A269217-5G
77001-084EA 43.5 USD
77001-084 77001-086 77001-098 77001-100 77001-102
2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide 97%
2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO
MW: 211.6880 g/mol
MDL Number: MFCD00267498
CAS Number: 32428-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR