6-Methoxypyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A269053-1G A269053-250MG A269053-5G A269053-100MG
77014-390EA 182.48 USD
77014-390 77014-392 77014-394 77683-821
6-Methoxypyrimidine-2,4(1H,3H)-dione 98%
6-Methoxypyrimidine-2,4(1H,3H)-dione

Precaución: For research use only.

Formula: C₅H₆N₂O₃
MW: 142.11 g/mol
MDL Number: MFCD00475644
CAS Number: 29458-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR