Ethyl 4-bromo-3-methoxybenzoate 95%

Supplier: Ambeed
Warning

A268896-1G A268896-250MG
77014-376EA 27.59 USD
77014-376 77014-378
Ethyl 4-bromo-3-methoxybenzoate 95%
Ethyl 4-bromo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.09 g/mol
MDL Number: MFCD14698173
CAS Number: 933585-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR