Methyl-2,5-difluoro-4-nitrobenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2,5-difluoro-4-nitrobenzoate
A268558-10G
A268558-1G
A268558-250MG
A268558-5G
77001-068EA
381.93
USD
77001-068
77001-070
77001-072
77672-384
Methyl-2,5-difluoro-4-nitrobenzoate 95%
Methyl 2,5-difluoro-4-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₈H₅F₂NO₄ MW: 217.13 g/mol |
MDL Number:
MFCD08689670 CAS Number: 924868-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...