4-Bromo-2,3-dihydro-1H-inden-2-ol 97%

Supplier: Ambeed
Warning

A268335-100MG A268335-250MG
77014-366EA 88.06 USD
77014-366 77014-368
4-Bromo-2,3-dihydro-1H-inden-2-ol 97%
4-Bromo-2,3-dihydro-1H-inden-2-ol

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD23381098
CAS Number: 125141-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR