Methyl-3,4,5-tris(benzyloxy)benzoate 95%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Tribenzyloxybenzoic Acid Methyl Ester, Methyl 3,4,5-tris(benzyloxy)benzoate, 3,4,5-Tris(benzyloxy)benzoic Acid Methyl Ester, Methyl 3,4,5-Tribenzyloxybenzoate

A268071-100G A268071-10G A268071-1G A268071-250MG A268071-25G A268071-5G
77014-354EA 228.1 USD
77014-354 77014-356 77014-358 77014-360 77014-362 77014-364
Methyl-3,4,5-tris(benzyloxy)benzoate 95%
Methyl-3,4,5-tris(benzyloxy)benzoate

Precaución: for Research Use Only

Formula: C₂₉H₂₆O₅
MW: 454.52 g/mol
MDL Number: MFCD03425302
CAS Number: 70424-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR