N-Methyl-4-(dimethylamino)benzylamine Hydrochloride 97%

Supplier: Ambeed
Warning

A268005-1G A268005-250MG
77014-350EA 60 USD
77014-350 77014-352
N-Methyl-4-(dimethylamino)benzylamine Hydrochloride 97%
N-Methyl-4-(dimethylamino)benzylamine Hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MDL Number: MFCD07106762
CAS Number: 1158441-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR