4-(4-Bromophenyl)dibenzofuran 96%

Supplier: Ambeed
Warning

A267985-100G A267985-100MG A267985-1G A267985-250MG A267985-25G A267985-5G
77014-338EA 259 USD
77014-338 77014-340 77014-342 77014-344 77014-346 77014-348
4-(4-Bromophenyl)dibenzofuran 96%
4-(4-Bromophenyl)dibenzofuran

Precaución: for Research Use Only

Formula: C₁₈H₁₁BrO
MDL Number: MFCD19441306
CAS Number: 955959-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR