2,5-Dimethoxypyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A267814-100G A267814-10G A267814-25G A267814-5G
77014-330EA 35 USD
77014-330 77014-332 77014-334 77014-336
2,5-Dimethoxypyrimidin-4-amine 95%
2,5-Dimethoxypyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MDL Number: MFCD20486065
CAS Number: 6960-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR