4-Bromoisatin 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromoindoline-2,3-dione

A267730-100G A267730-1G A267730-25G A267730-500G A267730-5G A267730-10G
77001-052EA 103.97 USD
77001-052 77001-054 77001-056 77001-058 77001-060 77601-150
4-Bromoisatin 98%
4-Bromoisatin

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MW: 226.0269 g/mol
MDL Number: MFCD03618555
CAS Number: 20780-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR