tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate 96%

Supplier: Ambeed
Warning

A267542-100MG A267542-1G A267542-250MG A267542-5G
77014-320EA 19 USD
77014-320 77014-322 77014-324 77014-326
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate 96%
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂S
MDL Number: MFCD03659715
CAS Number: 215183-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR