2-Oxoglutaric acid 95%

Supplier: Ambeed
Danger

Synonyms: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid

A267541-100G A267541-25G
77001-048EA 12.74 USD
77001-048 77304-978
2-Oxoglutaric acid 95%
2-Oxoglutaric acid

Precaución: for Research Use Only

Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
MDL Number: MFCD00004165
CAS Number: 328-50-7
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR