Tetraphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

A267236-1000G A267236-100G A267236-10G A267236-25G A267236-500G
77031-868EA 78 USD
77031-868 77031-870 77031-872 77031-874 77031-876
Tetraphenylphosphonium bromide 98%
Tetraphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₄H₂₀BrP
MW: 419.3 g/mol
MDL Number: MFCD00011915
CAS Number: 2751-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR