4-(5-Bromopyrimidin-2-yl)thiomorpholine 97%

Supplier: Ambeed
Warning

A267209-1G A267209-250MG A267209-5G
77014-312EA 24.41 USD
77014-312 77014-314 77014-316
4-(5-Bromopyrimidin-2-yl)thiomorpholine 97%
4-(5-Bromopyrimidin-2-yl)thiomorpholine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN₃S
MW: 260.15 g/mol
MDL Number: MFCD09909661
CAS Number: 1341761-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR