1-(4-Bromophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one 95%

Supplier: Ambeed
Warning

A266835-10G A266835-250MG
76988-256EA 472.11 USD
76988-256 76988-258
1-(4-Bromophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one 95%
1-(4-Bromophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one

Precaución: for Research Use Only

Formula: C₁₁H₇BrN₄O
MW: 291.10 g/mol
MDL Number: MFCD06661809
CAS Number: 864872-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR