(S)-(-)-N-(1-Phenylethyl)phthalamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-N-(alpha-Methylbenzyl)phthalamic acid, (S)-(-)-N-(1-Phenylethyl)phthalamic acid, (S)-2-((1-Phenylethyl)carbamoyl)benzoic acid

A265974-100MG A265974-1G A265974-250MG A265974-5G
77031-850EA 18.04 USD
77031-850 77031-852 77031-854 77031-856
(S)-(-)-N-(1-Phenylethyl)phthalamic acid 98%
(S)-(-)-N-(1-Phenylethyl)phthalamic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃
MW: 269.3 g/mol
MDL Number: MFCD00013979
CAS Number: 21752-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR