3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%

Supplier: Ambeed
Warning

Synonyms: Piperidine-3-carboxaldehyde, N-BOC protected, tert-Butyl 3-formylpiperidine-1-carboxylate

A265676-1G A265676-25G A265676-5G A265676-10G
77014-282EA 10.61 USD
77014-282 77014-284 77568-524 77683-814
3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%
3-Formyl-N-(tert-butoxycarbonyl)piperidine

Precaución: For research use only.

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
MDL Number: MFCD02179020
CAS Number: 118156-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR