2-Acrylamido-2-methylpropanesulfonic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Acrylamido-2-methyl-1-propanesulphonic acid

A265646-25G A265646-100G
77001-028EA 5.31 USD
77001-028 77593-646
2-Acrylamido-2-methylpropanesulfonic acid 98%
2-Acrylamido-2-methylpropanesulphonic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄S
MW: 207.25 g/mol
MDL Number: MFCD00007522
CAS Number: 15214-89-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR