6,11-Dihydrodibenzo[b,e]thiepin-11-one 98%

Supplier: Ambeed
Warning

A265211-25G A265211-5G A265211-100G
77001-014EA 47.75 USD
77001-014 77001-016 77672-372
6,11-Dihydrodibenzo[b,e]thiepin-11-one 98%
6,11-Dihydrodibenzo[b,e]thiepin-11-one

Precaución: for Research Use Only

Formula: C₁₄H₁₀OS
MW: 226.29 g/mol
MDL Number: MFCD02093738
CAS Number: 1531-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR