H-D-ORN(boc)-OH 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: H-D-Orn(Boc)-OH

A265139-10G A265139-1G A265139-250MG A265139-5G A265139-25G
76988-238EA 177 USD
76988-238 76988-240 76988-242 77571-648 77683-812
H-D-ORN(boc)-OH 97%, Ambeed.Inc
H-D-ORN(boc)-OH

Precaución: For research use only.

Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
MDL Number: MFCD00237206
CAS Number: 184576-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR