6-Bromo-5-fluoropyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-fluoropicolinic acid

A265037-100MG A265037-10G A265037-1G A265037-250MG A265037-5G
77031-826EA 5 USD
77031-826 77031-828 77031-830 77031-832 77031-834
6-Bromo-5-fluoropyridine-2-carboxylic acid 98%
6-Bromo-5-fluoropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD13185797
CAS Number: 1052714-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR