1-(3,4-Dihydroxyphenyl)pentan-1-one 95%

Supplier: Ambeed
Warning

A264979-100MG A264979-1G A264979-250MG
77014-262EA 21.22 USD
77014-262 77014-264 77014-266
1-(3,4-Dihydroxyphenyl)pentan-1-one 95%
1-(3,4-Dihydroxyphenyl)pentan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD09031813
CAS Number: 2525-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR