(±)-3-Chloromandelic acid 98%

Supplier: Ambeed
Warning

Synonyms: (±)-3-Chlorophenylglycolic acid, 3-Chloromandelic acid

A264327-100G A264327-1G A264327-25G A264327-5G
77001-004EA 227.04 USD
77001-004 77001-006 77001-008 77001-010
(±)-3-Chloromandelic acid 98%
(±)-3-Chloromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00829409
CAS Number: 16273-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR