8-(Benzyloxy)-2-chloroquinoline 98%

Supplier: Ambeed
Warning

A264315-100MG A264315-1G A264315-250MG A264315-5G
77014-240EA 14.86 USD
77014-240 77014-242 77014-244 77014-246
8-(Benzyloxy)-2-chloroquinoline 98%
8-(Benzyloxy)-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₁₆H₁₂ClNO
MW: 269.72 g/mol
MDL Number: MFCD09261075
CAS Number: 343788-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR