1,3-Dimesitylimidazol-2-ylidene 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, IMes, 1,3-Bis(2,4,6-trimethylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene

A264210-1G A264210-250MG A264210-25G
76988-232EA 80 USD
76988-232 76988-234 77568-516
1,3-Dimesitylimidazol-2-ylidene 98%
1,3-Dimesitylimidazol-2-ylidene

Precaución: For research use only.

Formula: C₂₁H₂₄N₂
MDL Number: MFCD09265338
CAS Number: 141556-42-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR