3-Bromo-6,7-dihydro-5H-pyrrolo[3,4-b]pyridine hydrochloride 95%

Supplier: Ambeed
Warning

A263659-100MG A263659-1G
76988-230EA 88.06 USD
76988-230 77571-632
3-Bromo-6,7-dihydro-5H-pyrrolo[3,4-b]pyridine hydrochloride 95%
3-Bromo-6,7-dihydro-5H-pyrrolo[3,4-b]pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈BrClN₂
MDL Number: MFCD19688582
CAS Number: 1394117-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR